siRNA Details
| Gene Symbol: Ccdc27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191063.1 | NP_001177992.1 | 9, 10 | 1432 | ||
| XM_008764384.1 | XP_008762606.1 | 10, 11 | 1596 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc27| Gene Symbol: Ccdc27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191063.1 | NP_001177992.1 | 9, 10 | 1432 | ||
| XM_008764384.1 | XP_008762606.1 | 10, 11 | 1596 | |||