siRNA Details
| Gene Symbol: Olr1875 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002728787.3 | XP_002728833.2 | 1 | 252 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr1875| Gene Symbol: Olr1875 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002728787.3 | XP_002728833.2 | 1 | 252 | ||
Set Membership: |
|