siRNA Details
| Gene Symbol: LOC687088 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760293.1 | XP_008758515.1 | 1 | 631 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC687088| Gene Symbol: LOC687088 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760293.1 | XP_008758515.1 | 1 | 631 | ||