siRNA Details
| Gene Symbol: Olr702 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_230251.6 | XP_230251.6 | 1 | 717 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr702| Gene Symbol: Olr702 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_230251.6 | XP_230251.6 | 1 | 717 | ||