siRNA Details
| Gene Symbol: Zcwpw2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008766697.1 | XP_008764919.1 | 6, 7 | 1655 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zcwpw2| Gene Symbol: Zcwpw2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008766697.1 | XP_008764919.1 | 6, 7 | 1655 | ||