siRNA Details
| Gene Symbol: Ric3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001115045.3 | NP_001108517.1 | 6 | 889 | ||
| NM_001277069.1 | NP_001263998.1 | 6 | 886 | |||
| GenBank mRNA | AM422212.1 | - | 6 | 886 | ||
| AM422213.1 | - | 6 | 889 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ric3| Gene Symbol: Ric3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001115045.3 | NP_001108517.1 | 6 | 889 | ||
| NM_001277069.1 | NP_001263998.1 | 6 | 886 | |||
| GenBank mRNA | AM422212.1 | - | 6 | 886 | ||
| AM422213.1 | - | 6 | 889 | |||