siRNA Details
| Gene Symbol: Stk33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759823.1 | XP_008758045.1 | 4 | 866 | ||
| XM_008759824.1 | XP_008758046.1 | 3 | 571 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stk33| Gene Symbol: Stk33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759823.1 | XP_008758045.1 | 4 | 866 | ||
| XM_008759824.1 | XP_008758046.1 | 3 | 571 | |||