siRNA Details
| Gene Symbol: Lrr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109483.1 | NP_001102953.1 | 3 | 813 | ||
| XM_008764719.1 | XP_008762941.1 | 2 | 831 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrr1| Gene Symbol: Lrr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109483.1 | NP_001102953.1 | 3 | 813 | ||
| XM_008764719.1 | XP_008762941.1 | 2 | 831 | |||