siRNA Details
| Gene Symbol: LOC691141 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109624.1 | NP_001103094.1 | 5, 6 | 655 | ||
| XM_006242674.2 | XP_006242736.1 | 4, 5 | 1893 | |||
| XM_008765985.1 | XP_008764207.1 | 6, 7 | 1953 | |||
| XM_008765986.1 | XP_008764208.1 | 6, 7 | 1953 | |||
| GenBank mRNA | FQ227025.1 | - | 4, 5 | 541 | ||
| FQ230556.1 | - | 5, 6 | 640 | |||

LOC691141
Genomic Map