siRNA Details
| Gene Symbol: Zfpm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242627.1 | NP_001229556.1 | 4 | 448 | ||
| XM_008772646.1 | XP_008770868.1 | 3 | 3916 | |||
| XM_008772647.1 | XP_008770869.1 | 3 | 3916 | |||
| XM_008772649.1 | XP_008770871.1 | 1 | 58 | |||

Zfpm1
Genomic Map