siRNA Details
| Gene Symbol: RGP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080496.2 | NP_001073965.2 | 4 | 448 | ||
| GenBank mRNA | AK289566.1 | - | 4 | 441 | ||
| BC001725.1 | - | 4 | 376 | 3533857 | ||
| D87447.1 | - | 4 | 385 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGP1| Gene Symbol: RGP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080496.2 | NP_001073965.2 | 4 | 448 | ||
| GenBank mRNA | AK289566.1 | - | 4 | 441 | ||
| BC001725.1 | - | 4 | 376 | 3533857 | ||
| D87447.1 | - | 4 | 385 | |||