siRNA Details
| Gene Symbol: Amph | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022217.1 | NP_071553.1 | 9 | 731 | ||
| GenBank mRNA | Y13381.1 | - | 9 | 731 | ||
| Gene Symbol: LOC100909679 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006257729.2 | XP_006257791.1 | 2 | 867 | ||
| Gene Symbol: LOC100910792 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008771838.1 | XP_008770060.1 | 8 | 674 | ||

Amph
Genomic Map