siRNA Details
| Gene Symbol: Smdt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_235517.6 | XP_235517.6 | 3 | 687 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smdt1| Gene Symbol: Smdt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_235517.6 | XP_235517.6 | 3 | 687 | ||