siRNA Details
Gene Symbol: Smdt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_235517.6 | XP_235517.6 | 2 | 436 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Smdt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_235517.6 | XP_235517.6 | 2 | 436 |