siRNA Details
| Gene Symbol: NEWGENE_1595998 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008767890.1 | XP_008766112.1 | 4 | 1279 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NEWGENE_1595998| Gene Symbol: NEWGENE_1595998 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008767890.1 | XP_008766112.1 | 4 | 1279 | ||