siRNA Details
| Gene Symbol: Adat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109611.1 | NP_001103081.1 | 9 | 1452 | ||
| XM_006255634.2 | XP_006255696.1 | 9 | 2292 | |||
| XM_006255635.2 | XP_006255697.1 | 9 | 2418 | |||
| XM_006255636.2 | XP_006255698.1 | 9 | 2246 | |||
| XM_006255638.1 | XP_006255700.1 | 8 | 1297 | |||
| XM_008772535.1 | XP_008770757.1 | 9 | 2292 | |||

Adat1
Genomic Map