siRNA Details
| Gene Symbol: DNAJC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256864.1 | NP_001243793.1 | 16 | 2635 | ||
| NM_001256865.1 | NP_001243794.1 | 17 | 2583 | |||
| NM_014787.3 | NP_055602.1 | 16 | 2470 | |||
| GenBank mRNA | AB007942.1 | - | 16 | 2410 | ||
| AK296408.1 | - | 17 | 2537 | |||
| BC051722.1 | - | 17 | 2274 | |||
| BC109279.2 | - | 16 | 2568 | 40028812 | ||
| BC109280.1 | - | 16 | 2571 | 40028814 | ||
| BX647203.1 | - | 17 | 2624 | |||

DNAJC6
Genomic Map