siRNA Details
| Gene Symbol: RGD1561671 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_341496.7 | XP_341497.4 | 1 | 192 | ||
| Gene Symbol: Uqcc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108528.1 | NP_001101998.1 | 2 | 233 | ||
| XM_006256124.2 | XP_006256186.1 | 3 | 258 | |||
| XM_008772733.1 | XP_008770955.1 | 3 | 682 | |||
| GenBank mRNA | BC169127.1 | - | 400 | 6918056 | ||

RGD1561671
Genomic Map