siRNA Details
| Gene Symbol: LOC100360638 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759212.1 | XP_008757434.1 | 5 | 1015 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100360638| Gene Symbol: LOC100360638 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759212.1 | XP_008757434.1 | 5 | 1015 | ||