siRNA Details
| Gene Symbol: LOC100360638 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759212.1 | XP_008757434.1 | 5 | 442 | ||
| Gene Symbol: Zfp780b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006228592.2 | XP_006228654.1 | 5 | 1847 | ||
| XM_006228593.2 | XP_006228655.1 | 6 | 780 | |||
| XM_006228595.2 | XP_006228657.1 | 6 | 690 | |||
| XM_006228596.2 | XP_006228658.1 | 6 | 689 | |||
| XM_006228597.2 | XP_006228659.1 | 5 | 525 | |||
| XM_006228598.2 | XP_006228660.1 | 5 | 699 | |||
| XM_006228600.2 | XP_006228662.1 | 3 | 617 | |||
| XM_008759204.1 | XP_008757426.1 | 6 | 599 | |||
| XM_008759205.1 | XP_008757427.1 | 5 | 1841 | |||
| XM_008759206.1 | XP_008757428.1 | 5 | 693 | |||

LOC100360638
Genomic Map