siRNA Details
Gene Symbol: Zfp780b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006228592.2 | XP_006228654.1 | 5 | 1826 | ||
XM_006228593.2 | XP_006228655.1 | 6 | 759 | |||
XM_006228595.2 | XP_006228657.1 | 6 | 669 | |||
XM_006228596.2 | XP_006228658.1 | 6 | 668 | |||
XM_006228597.2 | XP_006228659.1 | 5 | 504 | |||
XM_006228598.2 | XP_006228660.1 | 5 | 678 | |||
XM_006228600.2 | XP_006228662.1 | 3 | 596 | |||
XM_008759204.1 | XP_008757426.1 | 6 | 578 | |||
XM_008759205.1 | XP_008757427.1 | 5 | 1820 | |||
XM_008759206.1 | XP_008757428.1 | 5 | 672 |
Gene Symbol: LOC100360638 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759212.1 | XP_008757434.1 | 5 | 421 |
