siRNA Details
| Gene Symbol: LOC102553760 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759210.1 | XP_008757432.1 | 4 | 2635 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102553760| Gene Symbol: LOC102553760 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759210.1 | XP_008757432.1 | 4 | 2635 | ||