siRNA Details
| Gene Symbol: Tvp23b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107568.3 | NP_001101038.2 | 3, 4 | 284 | ||
| GenBank mRNA | BC168178.1 | - | 3, 4 | 282 | ||
| CK483802.1 | - | 3, 4 | 284 | |||
| FQ212667.1 | - | 3, 4 | 320 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tvp23b| Gene Symbol: Tvp23b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107568.3 | NP_001101038.2 | 3, 4 | 284 | ||
| GenBank mRNA | BC168178.1 | - | 3, 4 | 282 | ||
| CK483802.1 | - | 3, 4 | 284 | |||
| FQ212667.1 | - | 3, 4 | 320 | |||