siRNA Details
| Gene Symbol: JAKMIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270934.1 | NP_001257863.1 | 12 | 2178 | ||
| NM_001270941.1 | NP_001257870.1 | 13 | 2241 | |||
| NM_001282282.1 | NP_001269211.1 | 12 | 1964 | |||
| NM_014790.4 | NP_055605.2 | 13 | 2241 | |||
| XM_017010097.1 | XP_016865586.1 | 14 | 2235 | |||
| XM_017010098.1 | XP_016865587.1 | 12 | 2121 | |||
| GenBank mRNA | AB011127.1 | - | 12 | 1872 | ||
| AF273057.1 | - | 13 | 2082 | |||
| AK291280.1 | - | 13 | 1963 | |||
| AK299724.1 | - | 12 | 1890 | |||
| BC017354.2 | - | 13 | 2006 | 5020741 | ||
| EF512550.1 | - | 12 | 1700 | |||

JAKMIP2
Genomic Map