siRNA Details
| Gene Symbol: Tmem116 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159625.1 | NP_001153097.1 | 2, 3 | 82 | ||
| XM_006249404.2 | XP_006249466.1 | 2, 3 | 432 | |||
| XM_006249406.2 | XP_006249468.1 | 2, 3 | 443 | |||
| GenBank mRNA | BC167104.1 | - | 2, 3 | 82 | ||

Tmem116
Genomic Map