siRNA Details
Gene Symbol: Harbi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113793.3 | NP_001107265.2 | 2 | 665 | ||
XM_006234587.2 | XP_006234649.1 | 2 | 1312 | |||
XM_006234588.2 | XP_006234650.1 | 2 | 717 | |||
GenBank mRNA | BC158734.1 | - | 2 | 665 | 9098654 |
