siRNA Details
| Gene Symbol: Prdm11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271530.1 | NP_001258459.1 | 8 | 3292 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prdm11| Gene Symbol: Prdm11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271530.1 | NP_001258459.1 | 8 | 3292 | ||