siRNA Details
| Gene Symbol: Dtx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751193.3 | XP_003751241.1 | 9 | 2468 | ||
| XM_006249412.2 | XP_006249474.1 | 9 | 2480 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dtx1| Gene Symbol: Dtx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751193.3 | XP_003751241.1 | 9 | 2468 | ||
| XM_006249412.2 | XP_006249474.1 | 9 | 2480 | |||