siRNA Details
| Gene Symbol: Gpr133 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001070157.6 | XP_001070157.3 | 22 | 2796 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpr133| Gene Symbol: Gpr133 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001070157.6 | XP_001070157.3 | 22 | 2796 | ||
Set Membership: |
|