siRNA Details
| Gene Symbol: Wdr87 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759284.1 | XP_008757506.1 | 13 | 6586 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wdr87| Gene Symbol: Wdr87 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759284.1 | XP_008757506.1 | 13 | 6586 | ||