siRNA Details
| Gene Symbol: Muc15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006234695.2 | XP_006234757.1 | 5 | 1677 | ||
| XM_008762107.1 | XP_008760329.1 | 4 | 1454 | |||
| XM_008762108.1 | XP_008760330.1 | 4 | 1490 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Muc15| Gene Symbol: Muc15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006234695.2 | XP_006234757.1 | 5 | 1677 | ||
| XM_008762107.1 | XP_008760329.1 | 4 | 1454 | |||
| XM_008762108.1 | XP_008760330.1 | 4 | 1490 | |||