siRNA Details
| Gene Symbol: Ccdc167 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109548.1 | NP_001103018.1 | 3 | 215 | ||
| XM_006256231.2 | XP_006256293.1 | 3 | 238 | |||
| XM_006256232.1 | XP_006256294.1 | 3 | 243 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc167| Gene Symbol: Ccdc167 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109548.1 | NP_001103018.1 | 3 | 215 | ||
| XM_006256231.2 | XP_006256293.1 | 3 | 238 | |||
| XM_006256232.1 | XP_006256294.1 | 3 | 243 | |||