siRNA Details
Gene Symbol: LOC685067 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749409.3 | XP_003749457.1 | 11 | 1908 | ||
GenBank mRNA | FQ221477.1 | - | 1832 | |||
FQ222982.1 | - | 11 | 1815 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC685067 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749409.3 | XP_003749457.1 | 11 | 1908 | ||
GenBank mRNA | FQ221477.1 | - | 1832 | |||
FQ222982.1 | - | 11 | 1815 |