siRNA Details
Gene Symbol: Znf146 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759296.1 | XP_008757518.1 | 1 | 451 |
Gene Symbol: LOC102555183 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006247118.2 | XP_006247180.1 | 2 | 1271 |
