siRNA Details
| Gene Symbol: Iqck | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006230149.2 | XP_006230211.1 | 1, 2 | 204 | ||
| XM_006230150.1 | XP_006230212.1 | 1, 2 | 204 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Iqck| Gene Symbol: Iqck | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006230149.2 | XP_006230211.1 | 1, 2 | 204 | ||
| XM_006230150.1 | XP_006230212.1 | 1, 2 | 204 | |||