siRNA Details
| Gene Symbol: Anks4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127650.1 | NP_001121122.1 | 1 | 609 | ||
| GenBank mRNA | BC166493.1 | - | 1 | 609 | 7110362 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Anks4b| Gene Symbol: Anks4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127650.1 | NP_001121122.1 | 1 | 609 | ||
| GenBank mRNA | BC166493.1 | - | 1 | 609 | 7110362 | |