siRNA Details
| Gene Symbol: Ccdc62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134766.1 | NP_001128238.1 | 9 | 1719 | ||
| XM_006249355.2 | XP_006249417.1 | 9 | 2587 | |||
| XM_006249356.2 | XP_006249418.1 | 9 | 2533 | |||
| XM_006249357.2 | XP_006249419.1 | 9 | 2587 | |||
| XM_006249358.1 | XP_006249420.1 | 9 | 1660 | |||
| XM_008769265.1 | XP_008767487.1 | 8 | 1472 | |||
| GenBank mRNA | BC086549.1 | - | 5 | 896 | ||
| BC099139.1 | - | 5 | 998 | |||

Ccdc62
Genomic Map