siRNA Details
| Gene Symbol: Denr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001115046.2 | NP_001108518.1 | 5 | 299 | ||
| XM_008769260.1 | XP_008767482.1 | 5 | 350 | |||
| GenBank mRNA | BC158753.1 | - | 5 | 299 | 9102903 | |
| FQ210696.1 | - | 5 | 321 | |||
| FQ212845.1 | - | 5 | 319 | |||
| FQ220357.1 | - | 5 | 356 | |||
| FQ232826.1 | - | 5 | 320 | |||

Denr
Genomic Map