siRNA Details
| Gene Symbol: Gpr81 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145334.1 | NP_001138806.1 | 1 | 172 | ||
| GenBank mRNA | EU809461.1 | - | 1 | 172 | ||
| EU809462.1 | - | 1 | 148 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpr81| Gene Symbol: Gpr81 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145334.1 | NP_001138806.1 | 1 | 172 | ||
| GenBank mRNA | EU809461.1 | - | 1 | 172 | ||
| EU809462.1 | - | 1 | 148 | |||