siRNA Details
| Gene Symbol: U2af1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044291.1 | NP_001037756.1 | 2 | 153 | ||
| XM_006256228.2 | XP_006256290.1 | 2 | 170 | |||
| GenBank mRNA | BC105905.1 | - | 2 | 153 | 7374686 | |
| FQ229339.1 | - | 2 | 147 | |||

U2af1
Genomic Map