siRNA Details
Gene Symbol: Il17c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191115.1 | NP_001178044.1 | 2 | 261 | ||
XM_008772650.1 | XP_008770872.1 | 2 | 285 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Il17c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191115.1 | NP_001178044.1 | 2 | 261 | ||
XM_008772650.1 | XP_008770872.1 | 2 | 285 |