siRNA Details
| Gene Symbol: Il17c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191115.1 | NP_001178044.1 | 3 | 543 | ||
| XM_008772650.1 | XP_008770872.1 | 3 | 567 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Il17c| Gene Symbol: Il17c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191115.1 | NP_001178044.1 | 3 | 543 | ||
| XM_008772650.1 | XP_008770872.1 | 3 | 567 | |||