siRNA Details
| Gene Symbol: LOC690085 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008769276.1 | XP_008767498.1 | 6 | 521 | ||
| GenBank mRNA | AY724520.1 | - | 5 | 599 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC690085| Gene Symbol: LOC690085 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008769276.1 | XP_008767498.1 | 6 | 521 | ||
| GenBank mRNA | AY724520.1 | - | 5 | 599 | ||