siRNA Details
Gene Symbol: Pwp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168653.1 | NP_001162124.1 | 2 | 93 | ||
XM_006256257.2 | XP_006256319.1 | 2 | 704 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pwp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168653.1 | NP_001162124.1 | 2 | 93 | ||
XM_006256257.2 | XP_006256319.1 | 2 | 704 |