siRNA Details
| Gene Symbol: LOC100909893 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003748831.3 | XP_003748879.1 | 2 | 1103 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100909893| Gene Symbol: LOC100909893 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003748831.3 | XP_003748879.1 | 2 | 1103 | ||