siRNA Details
| Gene Symbol: NEWGENE_1565505 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760350.1 | XP_008758572.1 | 1, 2 | 653 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NEWGENE_1565505| Gene Symbol: NEWGENE_1565505 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760350.1 | XP_008758572.1 | 1, 2 | 653 | ||