siRNA Details
| Gene Symbol: Morn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006249367.2 | XP_006249429.1 | 3 | 567 | ||
| XM_008769278.1 | XP_008767500.1 | 3 | 542 | |||
| XM_008769279.1 | XP_008767501.1 | 4 | 597 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Morn3| Gene Symbol: Morn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006249367.2 | XP_006249429.1 | 3 | 567 | ||
| XM_008769278.1 | XP_008767500.1 | 3 | 542 | |||
| XM_008769279.1 | XP_008767501.1 | 4 | 597 | |||