siRNA Details
| Gene Symbol: Mipol1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750162.3 | XP_003750210.1 | 6, 7 | 767 | ||
| XM_008764676.1 | XP_008762898.1 | 6, 7 | 767 | |||
| XM_008764677.1 | XP_008762899.1 | 7, 8 | 1198 | |||
| XM_008764678.1 | XP_008762900.1 | 6, 7 | 767 | |||
| GenBank mRNA | FQ220776.1 | - | 1, 2 | 84 | ||

Mipol1
Genomic Map