siRNA Details
| Gene Symbol: Mipol1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750162.3 | XP_003750210.1 | 9 | 1031 | ||
| XM_008764676.1 | XP_008762898.1 | 9 | 1031 | |||
| XM_008764677.1 | XP_008762899.1 | 10 | 1462 | |||
| GenBank mRNA | FQ220776.1 | - | 4 | 348 | ||

Mipol1
Genomic Map