siRNA Details
| Gene Symbol: LOC691083 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144862.1 | NP_001138334.1 | 97 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC691083| Gene Symbol: LOC691083 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144862.1 | NP_001138334.1 | 97 | |||